Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBNA1BP2/EBNA1 Binding Protein 2/EBP2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569443
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9443 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9443 Supplier Bethyl Laboratories, Inc Supplier No. A305067AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

EBNA1BP2 is required for the processing of the 27S pre-rRNA.

Specifications

Antigen EBNA1BP2/EBNA1 Binding Protein 2/EBP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EBNA1BP2
Gene Accession No. Q99848
Gene Alias EBNA1-binding protein 2, EBP2, NOBP, nuclear FGF3 binding protein, nucleolar protein p40, nucleolar protein p40; homolog of yeast EBNA1-binding protein, P40
Gene Symbols EBNA1BP2
Host Species Rabbit
Immunogen Between 256 and 306
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10969
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.