Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBNA1BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555326 Supplier Invitrogen™ Supplier No. PA555326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESDESLVTDR ELQDAFSRGL LKPGLNVVLE GPKKAVNDVN GLKQCLAEFK RDLEWVERLD VTLGPVPEIG GSEAPAPQNK DQKAVDPEDD Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.

EBNA1BP2 is required for the processing of the 27S pre-rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBNA1BP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EBNA1BP2
Gene Accession No. Q99848
Gene Alias 1810014B19Rik; AA537058; B830003A16Rik; C81321; EBNA1 binding protein 2; EBNA1-binding protein 2; Ebna1bp2; EBP2; NOBP; nuclear FGF3 binding protein; nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; p40; Probable rRNA-processing protein EBP2
Gene Symbols EBNA1BP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EBNA1BP2. Recombinant protein control fragment (Product #RP-94115).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.