Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EBNA1BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100789 Supplier Invitrogen™ Supplier No. PA5100789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EBP2.

EBNA1BP2 is required for the processing of the 27S pre-rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBNA1BP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EBNA1BP2
Gene Accession No. Q99848, Q9D903
Gene Alias 1810014B19Rik; AA537058; B830003A16Rik; C81321; EBNA1 binding protein 2; EBNA1-binding protein 2; Ebna1bp2; EBP2; NOBP; nuclear FGF3 binding protein; nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; p40; Probable rRNA-processing protein EBP2
Gene Symbols EBNA1BP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human EBNA1BP2(Accession Q99848), corresponding to amino acid residues M161-D211.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10969, 69072
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.