Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    EBNA2 Antibody (PE2), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250382MFV450
Description
EBNA2 Monoclonal antibody specifically detects EBNA2 in Human, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP), ChIP assay SequencingSpecifications
| EBNA2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Recombinant fusion protein against EBNA2. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 | 
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, ChIP Assay | |
| PE2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Virus | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            