Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ebola Virus NP (subtype Sudan, strain Zaire) Monoclonal Antibody (2601)
GREENER_CHOICE

Catalog No. PIMA533250
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533250 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533250 Supplier Thermo Scientific Supplier No. MA533250
Add to Cart
Add to Cart

Ebola Virus NP (subtype Sudan, strain Zaire) Monoclonal Antibody (2601)

Ebola Virus NP (subtype Sudan, strain Zaire) Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ebola Virus NP (subtype Sudan, strain Zaire)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2601
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias EVD
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.