Learn More
Invitrogen™ Ebola Virus VP30 Polyclonal Antibody

Description
Ebola Virus VP30 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | Ebola Virus VP30 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.92 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene Alias | Ebola Virus |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of Ebola virus VP30 protein (Zaire ebolavirus (strain Mayinga-76)). The exact sequence is proprietary. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.