Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ebola Virus VP40 Monoclonal Antibody (F1-B4)
GREENER_CHOICE

Catalog No. PIMA544986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544986 Supplier Invitrogen™ Supplier No. MA544986
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Application notes: Indirect ELISA.

Ebola virus is an enveloped, negative-stranded RNA virus. This virus belongs to the family Filoviridae and has a distinctive morphology similar only to that of Marburg virus, but apparently has no antigenic characteristics of Marburg virus. Its genome is a nonsegmented RNA strand. SDS-PAGE profiles demonstrate the presence of seven structural proteins. These are the major nucleoprotein NP (104 K), glycoprotein GP (125K), L protein (180 K), and four proteins: VP40 (40 K), ViPX (35 K), VP30 (30 K) and VP24 (24 K). All the proteins are encoded by monocistronic mRNA transcripts complementary to virion RNA. GP is the major protein of the surface spikes and only this 125K viral protein is glycosylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ebola Virus VP40
Applications ELISA
Classification Monoclonal
Clone F1-B4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias EVD
Host Species Mouse
Immunogen Synthetic peptide within C-terminal Zaire ebolavirus VP40.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.