Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ebola Virus VP40 (subtype Bundibugyo, strain Uganda 2007) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581843 Supplier Invitrogen™ Supplier No. PA581843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40/Matrix protein VP40.

Ebola virus is an enveloped, negative-stranded RNA virus. This virus belongs to the family Filoviridae and has a distinctive morphology similar only to that of Marburg virus, but apparently has no antigenic characteristics of Marburg virus. Its genome is a nonsegmented RNA strand. SDS-PAGE profiles demonstrate the presence of seven structural proteins. These are the major nucleoprotein NP (104 K), glycoprotein GP (125K), L protein (180 K), and four proteins: VP40 (40 K), ViPX (35 K), VP30 (30 K) and VP24 (24 K). All the proteins are encoded by monocistronic mRNA transcripts complementary to virion RNA. GP is the major protein of the surface spikes and only this 125K viral protein is glycosylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ebola Virus VP40 (subtype Bundibugyo, strain Uganda 2007)
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias EVD
Host Species Rabbit
Immunogen Recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40/Matrix protein VP40 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.