Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV GP Antibody (02), DyLight 650 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305998C
Description
EBOV GP Monoclonal antibody specifically detects EBOV GP in Virus samples. It is validated for ELISASpecifications
EBOV GP | |
Monoclonal | |
DyLight 650 | |
second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Glycoprotein/ GP (AHX24649.1; Met1-Gln650). | |
0.1 mL | |
Primary | |
Virus | |
Purified |
ELISA | |
02 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
3160774 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction