Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV GP Antibody (06), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306458AF532
Description
EBOV GP Monoclonal antibody specifically detects EBOV GP in Virus samples. It is validated for ELISASpecifications
EBOV GP | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
3160774 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
06 | |
ELISA | |
second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Glycoprotein/ GP (AAC54887.1; Met1-Gln650). | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction