Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV GP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305820MFV500
Description
EBOV GP1 Polyclonal antibody specifically detects EBOV GP1 in Virus samples. It is validated for Western BlotSpecifications
EBOV GP1 | |
Polyclonal | |
Western Blot | |
second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
Produced in rabbits immunized with purified, recombinant EBOV GP1 (Accession#: ACI28624.1; Met1-Arg501). | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
3160774 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction