Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV VP40 protein Antibody (02), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312761R
Description
EBOV VP40 protein Monoclonal antibody specifically detects EBOV VP40 protein in Ebola virus samples. It is validated for Western Blot, ELISASpecifications
EBOV VP40 protein | |
Monoclonal | |
DyLight 550 | |
Mouse | |
Protein A purified | |
RUO | |
Ebola virus | |
Purified |
Western Blot, ELISA | |
02 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV VP40 protein (Accession#: YP_138522.1; Val31-Lys326). | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction