Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV VP40 protein Antibody (08), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP326102H
Description
EBOV VP40 protein Monoclonal antibody specifically detects EBOV VP40 protein in Virus samples. It is validated for Western Blot, ELISASpecifications
EBOV VP40 protein | |
Monoclonal | |
HRP | |
PBS | |
Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV VP40 protein Antibody, Mouse MAb (Accession#: YP_138522.1, Val31-Lys326) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
08 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction