Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ebselen

Catalog No. aaj63190ma
Click to view available options
Quantity:
10 mg

CAS: 60940-34-3 | C13H9NOSe | 274.192 g/mol

Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

Solubility
Soluble in ethanol, methanol, acetone, acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 60940-34-3
Melting Point 180°C to 181°C
Color White
Molecular Formula C13H9NOSe
Quantity 10 mg
MDL Number MFCD00210937
UN Number UN3283
Merck Index 14,3486
Synonym ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
Solubility Information Soluble in ethanol,methanol,acetone,acetonitrile.
InChI Key DYEFUKCXAQOFHX-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2
IUPAC Name 2-phenyl-1,2-benzoselenazol-3-one
Molecular Weight (g/mol) 274.192
PubChem CID 3194
ChEBI CHEBI:77543
Formula Weight 274.2
Physical Form Powder
Chemical Name or Material Ebselen
Show More Show Less
Hazard Category H330-H373
Hazard Statement GHS H Statement
H331-H373
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P271-P284-P304+P340-P310-P314-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
RTECSNumber DE4140750
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.