Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBV Early Antigens Antibody (1108-1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234516IR
Description
EBV Early Antigens Monoclonal specifically detects EBV Early Antigens in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
EBV Early Antigens | |
Monoclonal | |
DyLight 755 | |
Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like | |
Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
1108-1 | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Mouse | |
Protein A or G purified | |
RUO | |
Epstein-Barr virus (EBV), also designated human herpesvirus 4 (HHV-4), is a member of the herpesvirus family and is one of the most common human viruses. EBV infects B cells and, though often asymptomatic, it can cause infectious mononucleosis, a disease characterized by fatigue, fever, sore throat and muscle soreness. The EBV-induced early antigens (Ea) are among several antigen complexes that have been identified in EBV-infected cells. The Ea complex is composed of diffuse (Ea-D) and restricted (Ea-R) components. The activity of Ea-D is suppressed during latent infection. BMRF1, the gene that encodes for Ea-D, is closely associated with the gene encoding for EBV DNA polymerase, and Ea-D is essential for the activity of this polymerase. Ea-D forms a complex with EBV DNase and, together, they may play a role in viral replication. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction