Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBV Latent Membrane Protein 1 Antibody (LMPO24), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250383R
Description
EBV Latent Membrane Protein 1 Monoclonal specifically detects EBV Latent Membrane Protein 1 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular).Specifications
EBV Latent Membrane Protein 1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified plasma membranes from EBV Latent Membrane Protein 1 expressing insect cells infected with a recombinant EBV Latent Membrane Protein 1 baculovirus | |
0.1 mL | |
Cancer, Cellular Signaling, Immunology, Virology Bacteria and Parasites | |
Niedobitek et al., 1995 (PMID: 7605996): “The LMPO24 clone demonstrates strong and specific reactivity in immunohistochemical staining of paraffin-embedded tissues”. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry | |
LMPO24 | |
Flow Cytometry, Immunohistochemistry, Flow (Intracellular) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction