Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBV LMP1 Mouse anti-Virus, Unconjugated, Clone: S12, MilliporeSigma™

Catalog No. MABF22482MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22482MI 25 μg
MABF22481MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22482MI Supplier MilliporeSigma Supplier No. MABF224825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBV LMP1 Monoclonal antibody specifically detects EBV LMP1 in Virus samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBV LMP1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S12
Conjugate Unconjugated
Gene Alias Latent membrane protein;p63
Gene Symbols LMP1, BNLF1
Host Species Mouse
Immunogen A fusion protein (pKH548) consisting of 189 amino acids from the C-terminus region of LMP1 coupled to beta-galactosidase.
Molecular Weight of Antigen ∼59 kDa observed; 41.98 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone S12 is a mouse monoclonal antibody that specifically detects Latent membrane protein 1 (LMP1) in Epstein-Barr virus transformed cells.
Target Species Virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.