Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ EC Broth with MUG Bottled Broth 9mL Vials

Catalog No. 10486162 Shop All GVS Products
Change view
Click to view available options
Quantity:
9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-486-162 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-486-162 Supplier GVS™ Supplier No. 10496709
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EC Broths with MUG for the detection of Escherichia coli in water, milk and food samples by fluorogenic means. MUG detects anaerogenic strain that may not be detected in the conventional procedure. Ready-to-use 9mL ampoule shortens prep time and minimizes possible contamination.

Description

  • The presence of fluorescence using a long-wave UV light source confirms the presence of Escherichia coli (E.Coli) and no further confirmation is required.
  • MUG detects anaerogenic strains, which may not be detected in the conventional procedure. Lactose is a source of energy.
  • Casein peptone provides additional nutrients. The mixture of bile salts is inhibiting for gram-positive bacteria, particularly bacilli and fecal streptococci. The substrate 4-methylumbelliferyl-b-D-glucuronide is hydrolyzed by an enzyme, b-glucuronidase, possessed by most Escherichia coli and a few strains of Salmonella, Shigella and Yersinia, to produce a fluorescent end product, 4-methylumbelliferone.

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 9 mL Portions, Reduce Risk of Cross-Contamination.
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Use with membrane filters to culture bacteria, yeasts, and molds.
  • For the detection of Escherichia coli in water and food samples by fluorogenic means.
  • MUG detects anaerogenic strain that may not be detected in the conventional procedure.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits.
This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Appearance Yellow Liquid
Broth Type EC Broth with MUG Bottled Broth
Cap Color Black
Certifications/Compliance Batch-specific quality certificate in each pack
Color Yellow Clear
Content And Storage 2°C to 8°C
Additive Lactose, Casein Peptone, Bile Salts, 4-methylumbelliferyl-b-D-glucuronide
Detects Escherichia coli in water, milk and food.
Format Liquid, Vial
Manufacturing Origin United States
Media Type Broth Tube
Product Type Broth Tube
Sterility Sterile
Test Type Microbial Detection
Fill Volume 9.0 mL ±0.5 mL
Formulation with lactose, casein peptone, and bile salts.
For Use With (Application) Detect coliforms and E. coli from food and water samples
Includes 20 Tubes
Incubation Conditions 44.5°C ±2°C
Incubation Time 24 hr.
Material Borosilicate Glass, Phenolic, Rubber
No. per Pack 20
Packaging Type Vial
Quantity 9 mL
Reading Options Fluorescence Using Long-wave UV Light
Dimensions (Dia. x H) 19 x 65 mm
Height (Metric) 65 mm
Diameter (Metric) 19 mm
Protein Source Casein
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.