Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ecat1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983009
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3009 Supplier Bioss Supplier No. BS13596R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the KHDC1 family, members of which contain an atypical KH domain that may not bind RNA like canonical KH domains. This gene is specifically expressed in the oocytes, and recent studies suggest that it may function as a regulator of genomic imprinting in the oocyte. Mutations in this gene are associated with recurrent biparental complete hydatidiform mole.

Specifications

Antigen Ecat1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KHDC3L
Gene Accession No. Q587J8
Gene Alias C6orf221; ECAT1; ES cell-associated transcript 1 protein; HYDM2; KH domain containing 3 like, subcortical maternal complex member; KH domain containing 3-like, subcortical maternal complex member; KHDC3L; KHDC3-like protein
Gene Symbols KHDC3L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ecat1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154288
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.