Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552298
Description
Immunogen sequence: DEDFECLDSD DDLDFETHEP GEEASLKGTL DNLKSYMAQM DQELAHTCIS KSFTTRNQVE PVSQTTDNNS DEEDSGTGES VMAPVDVDLN LVSNILESYS SQAGLAGPAS NLLQSMGVQL PDNTDHRPTS KP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 69%.
Regulator of p53/TP53 stability and function. Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP (PubMed:16849563, PubMed:23880345). May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes (PubMed:19919181, PubMed:9928932). Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins (PubMed:19640839). The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex (PubMed:26711270). May play a role in regulation of pre-mRNA splicing (PubMed:24722212). [UniProt].
Specifications
| ECD | |
| Polyclonal | |
| Unconjugated | |
| ECD | |
| 5730461K03Rik; Ecd; ecdysoneless cell cycle regulator; ecdysoneless homolog; ecdysoneless homolog (Drosophila); GCR2; HSGT1; Human suppressor of GCR two; protein ecdysoneless homolog; protein SGT1; protein SGT1 homolog; SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.08 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95905 | |
| ECD | |
| Recombinant protein corresponding to Human ECD. Recombinant protein control fragment (Product #RP-89339). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction