Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581948
Description
Immunogen sequence: IMDPKELDKV FNDYTAVPDL YFENAMRFFN FSWRVTADQL RKAPNRDQWS MTPPMVNAYY SPTKNEIVFP AGILQAPFYT RSSPKALNFG GIGVVVGHEL THAFDDQGRE YDKDGNLRPW WKNSSVEAFK R.
Endothelin-converting enzyme 1 (ECE1) is located on chromosome 1p36.12 and encodes a zinc metalloprotease involved in the maturation of endothelin peptides, which are critical for vascular tone regulation and blood pressure control. ECE1 processes precursor proteins into active endothelins, particularly endothelin-1, the most potent vasoconstrictor known. This enzyme is expressed in multiple tissues, including endothelial cells, smooth muscle cells, and the central nervous system, where it participates in diverse physiological processes such as vasoconstriction, cell proliferation, and hormone secretion. Beyond its cardiovascular functions, ECE1 is implicated in neural development and neural crest cell migration. Dysregulation of ECE1 activity or endothelin signaling is associated with several pathologies, including hypertension, heart failure, and cancer progression due to its role in controlling vascular dynamics. Research into ECE1 aims to explore its therapeutic potential in managing cardiovascular disorders and its broader implications in systemic pathophysiology.
Specifications
ECE1 | |
Polyclonal | |
Unconjugated | |
ECE1 | |
AW322500; Ece; ECE 1; ECE1; ECE-1; ECE-1a; ECE-1b; ECE-1d; endothelin converting enzyme 1; endothelin-converting enzyme 1; endothelin-converting enzyme-1a; endothelin-converting enzyme-1b; endothelin-converting enzyme-1d | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P42892 | |
ECE1 | |
Recombinant protein corresponding to Human ECE1. Recombinant protein control fragment (Product #RP-99357). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction