Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1777 Supplier Bioss Supplier No. BS1190R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECE-1 is involved in proteolytic processing of endothelin precursors to biologically active peptides.

Specifications

Antigen ECE1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ECE1
Gene Accession No. P42892
Gene Alias AW322500; ECE; ECE 1; Ece1; ECE-1; ECE-1a; ECE-1b; ECE-1d; endothelin converting enzyme 1; Endothelin-converting enzyme 1; endothelin-converting enzyme-1a; endothelin-converting enzyme-1b; endothelin-converting enzyme-1d
Gene Symbols ECE1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ECE1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1889
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.