Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100495
Description
Antibody detects endogenous levels of total ECE2.
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
Specifications
ECE2 | |
Polyclonal | |
Unconjugated | |
Ece2 | |
1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
107522, 408243, 9718 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
B2RQR8, P0DPD6 | |
Ece2 | |
A synthesized peptide derived from human ECE2(Accession P0DPD6), corresponding to amino acid residues P801-W811. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction