Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547175
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547175 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547175 Supplier Invitrogen™ Supplier No. PA547175
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human ECE-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECE2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ece2
Gene Accession No. P0DPD6
Gene Alias 1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740
Gene Symbols Ece2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ECE-2 Gly104-Trp787.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9718
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.