Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569351 Supplier Invitrogen™ Supplier No. PA569351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECE2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ece2
Gene Accession No. 0
Gene Alias 1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740
Gene Symbols Ece2
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat ECE2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408243
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.