Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECE2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1753R

Description
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.Specifications
ECE2 | |
Polyclonal | |
Unconjugated | |
Ece2 | |
1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; BOS_988; ECE2; ECE-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; Endothelin-converting enzyme 2; endothelin-converting enzyme 2 isoform 2a-1; endothelin-converting enzyme 2 isoform 2a-2; endothelin-converting enzyme 2 isoform 2b-1; endothelin-converting enzyme 2 isoform 2b-2; Endothelin-converting enzyme 2 region; KIAA0604; Methyltransferase-like region; UNQ403/PRO740 | |
Rabbit | |
Protein A | |
RUO | |
9718 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P0DPD6 | |
Ece2 | |
KLH conjugated synthetic peptide derived from human ECE2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction