Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECEL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | ECEL1 |
---|---|
Dilution | Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ECEL1 Polyclonal antibody specifically detects ECEL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ECEL1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.2, 40% glycerol | |
9427 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
damage induced neuronal endopeptidase, DINE, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.11, ECEX, endothelin converting enzyme-like 1, endothelin-converting enzyme-like 1, Xce protein, XCEX converting enzyme | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LFSLTVSLDDRNSSRYVIRIDQDGLTLPERTLYLAQDEDSEKILAAYRVFMERVLSLLGADAVEQKAQEILQVEQQLANITVSEHDDLR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title