Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114322 Supplier Invitrogen™ Supplier No. PA5114322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes ECH in Arabidopsis thaliana.

Mediates trans-Golgi-network trafficking and cell elongation. Required for keeping the appropriate balance between secretory trafficking and vacuolar targeting of a subset of proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ECH
Gene Accession No. Q8LEK2
Gene Alias AT1G09330; ECH; ECHIDNA; Golgi apparatus membrane protein-like protein ECHIDNA; golgi apparatus membrane protein-like protein ECHIDNA protein; T31J12.5; T31J12_5; unknown
Gene Symbols ECH
Host Species Rabbit
Immunogen Synthetic peptide within arath ECH aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 837454
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.