Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECH1 Antibody (002), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290288APC
Description
ECH1 Monoclonal antibody specifically detects ECH1 in Human samples. It is validated for Western Blot, ELISASpecifications
ECH1 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
002 | |
Western Blot, ELISA | |
delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial, delta3,5-delta2,4-dienoyl-CoA isomerase, dienoyl-CoA isomerase, EC 5.3.3.-, enoyl CoA hydratase 1, peroxisomal, enoyl Coenzyme A hydratase 1, peroxisomal, HPXEL, peroxisomal enoyl-CoA hydratase 1 | |
Rabbit immunized with purified, recombinant Human ECH1 (Uniprot#: Q13011, Thr34-Leu328) | |
0.1 mL | |
Lipid and Metabolism | |
1891 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction