Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582041
Description
Immunogen sequence: ISLRLTGSSA QEEASGVALG EAPDHSYESL RVTSAQKHVL HVQLNRPNKR NAMNKVFWRE MVECFNKISR DADCRAVVIS GAGKMFTAGI DLMDMASDIL QPKGDDVARI SWYLRDIITR YQETFNVIER CPK.
This gene encodes a member of the hydratase/isomerase superfamily. The gene product shows high sequence similarity to enoyl-coenzyme A (CoA) hydratases of several species, particularly within a conserved domain characteristic of these proteins. The encoded protein, which contains a C-terminal peroxisomal targeting sequence, localizes to the peroxisome. The rat ortholog, which localizes to the matrix of both the peroxisome and mitochondria, can isomerize 3-trans,5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA, indicating that it is a delta3,5-delta2,4-dienoyl-CoA isomerase. This enzyme functions in the auxiliary step of the fatty acid beta-oxidation pathway. Expression of the rat gene is induced by peroxisome proliferators.
Specifications
ECH1 | |
Polyclonal | |
Unconjugated | |
ECH1 | |
AA617331; delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial; delta3,5-delta2,4-dienoyl-CoA isomerase; delta3,5-delta2,4-dienoyl-coenzyme A isomerase; dienoyl-CoA isomerase; ECH1; enoyl CoA hydratase 1, peroxisomal; enoyl coenzyme A hydratase 1, peroxisomal; enoyl hydratase-like protein peroxisomal; enoyl hydratase-like protein, peroxisomal; enoyl-CoA hydratase 1; enoyl-CoA hydratase 1, peroxisomal; HPXEL; Peroxisomal enoyl hydratase-like protein; peroxisomal enoyl-CoA hydratase 1; peroxisomal/mitochondrial dienoyl-CoA isomerase; Pxel | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1891 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13011 | |
ECH1 | |
Recombinant protein corresponding to Human ECH1. Recombinant protein control fragment (Product #RP-101864). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction