Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHHADH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109679 Supplier Invitrogen™ Supplier No. PA5109679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EHHADH Polyclonal Antibody for Western Blot, ELISA

As an L-hydroxy-specific enzyme, EHHADH (enoyl-CoA-hydratase:3-hydroxyacyl-CoA dehydrogenase), also known as Peroxisomal L-bifunctional enzyme, is a 723 amino acid protein has an essential tripeptide sequence on its carboxyl-terminus that is required for peroxisomal transport. EHHADH-null mice only exhibit a blunted peroxisome proliferative response when challenged with a peroxisome proliferator. Since there were no observed changes in lipid metabolism, this evidence suggests that enoyl-CoAs were diverted to the D-hydroxy-specific beta-oxidation system for metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHHADH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EHHADH
Gene Accession No. P07896, Q08426, Q9DBM2
Gene Alias 1; 1300002P22Rik; 3,2-trans-enoyl-CoA isomerase; 3-hydroxyacyl-CoA dehydrogenase; ECHD; EHHADH; Enoyl-CoA hydratase/3,2-trans-enoyl-CoA isomerase; enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; FRTS3; HD; L-3-hydroxyacyl-CoA dehydrogenase; LBFP; L-bifunctional enzyme; L-bifunctional protein, peroxisomal; Lbp; L-PBE; L-specific bifunctional protein; L-specific multifunctional beta-oxdiation protein; MEF; Mfe; Mfe1; MFP; MFP1; multifunctional enzyme type 1; Pbe; PBFE; perMFE-1; Peroxisomal bifunctional enzyme; peroxisomal bifunctional enzyme type 1; peroxisomal enoyl-CoA hydratase
Gene Symbols EHHADH
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171142, 1962, 74147
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.