Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECHDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543232
Description
Peptide sequence: GVMMLQLLEK VIELENWTEG KGLIVRGAKN TFSSGSDLNA VKSLGTPEDG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 100%.
ECHDC1 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC1 remains unknown.
Specifications
| ECHDC1 | |
| Polyclonal | |
| Unconjugated | |
| ECHDC1 | |
| 1700028A24Rik; AI314462; AI930038; D10Ertd667e; dJ351K20.2; ECHDC1; enoyl CoA hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1 isoform 2; enoyl-CoA hydratase domain-containing 1-like protein; enoyl-CoA hydratase domain-containing protein 1; enoyl-CoA hydratase domain-containing protein-like protein; epididymis secretory protein Li 76; ethylmalonyl-CoA decarboxylase; ethylmalonyl-CoA decarboxylase 1; HEL-S-76; Methylmalonyl-CoA decarboxylase; MMCD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55862 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NTX5 | |
| ECHDC1 | |
| Synthetic peptide directed towards the middle region of human ECHDC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction