Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECHDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557272 Supplier Invitrogen™ Supplier No. PA557272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MALKQEMAKS LLKTASLSGR TKLLHQTGLS LYSTSHGFYE EEVKKTLQQF PGGSIDLQKE DNGIGILTLN NPSRMNAF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 71%.

ECHDC1 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECHDC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ECHDC1
Gene Accession No. Q9NTX5
Gene Alias 1700028A24Rik; AI314462; AI930038; D10Ertd667e; dJ351K20.2; ECHDC1; enoyl CoA hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1 isoform 2; enoyl-CoA hydratase domain-containing 1-like protein; enoyl-CoA hydratase domain-containing protein 1; enoyl-CoA hydratase domain-containing protein-like protein; epididymis secretory protein Li 76; ethylmalonyl-CoA decarboxylase; ethylmalonyl-CoA decarboxylase 1; HEL-S-76; Methylmalonyl-CoA decarboxylase; MMCD
Gene Symbols ECHDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ECHDC1. Recombinant protein control fragment (Product #RP-95051).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.