Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECHDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589924 Supplier Invitrogen™ Supplier No. PA589924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLRVLCLLRP WRPLRARGCA SDGAAGGSEI QVRALAGPDQ GITEILMNRP SARNALGNVF VSELLETLAQ LREDRQVRVL LFRSGVKGVF CAGADLKERE QMSEAEVGVF VQRLRGLMND IAAFPAPTIA AMDGFALGGG LELALACDLR VAASSAVMGL IETTRGLLPG AGGTQRLPRC LGVALAKELI FTGRRLSGTE AHVLGLVNHA VAQNEEGDAA YQRARALAQE ILPQAPIAVR LGKVAIDRGT EVDIASGMAI EGMCYAQNIP TRDRLEGMAA FREKRTPKFV GK; Positive Samples: Mouse stomach, Mouse thymus, Mouse heart, Rat lung; Cellular Location: Mitochondrion.

ECHDC2 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECHDC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Echdc2
Gene Accession No. Q3TLP5
Gene Alias 1300017C12Rik; 2610009M20Rik; D4Ertd765e; ECHDC2; enoyl CoA hydratase domain containing 2; enoyl Coenzyme A hydratase domain containing 2; enoyl-CoA hydratase domain containing 2; enoyl-CoA hydratase domain-containing protein 2, mitochondrial; hypothetical protein LOC513795; id:ibd1032; RGD1308525; Unknown (protein for MGC:85763); wu:fc83b02; zgc:56321; zgc:85763
Gene Symbols Echdc2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-292 of human ECHDC2 (NP_0011858901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298381, 52430
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.