Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECHDC2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982573
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2573 Supplier Bioss Supplier No. BS13049R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECHDC2 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC2 remains unknown.

Specifications

Antigen ECHDC2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Echdc2
Gene Accession No. Q86YB7
Gene Alias 1300017C12Rik; 2610009M20Rik; BOS_3529; D4Ertd765e; ECHDC2; ECHDC2 protein-like; enoyl CoA hydratase domain containing 2; enoyl Coenzyme A hydratase domain containing 2; enoyl-CoA hydratase domain containing 2; Enoyl-CoA hydratase domain-containing protein 2, mitochondrial; id:ibd1032; LOC100347669; RGD1308525; Unknown (protein for MGC:85763); wu:fc83b02; zgc:56321; zgc:85763
Gene Symbols Echdc2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ECHDC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55268
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.