Learn More
Torrey Pines Scientific™ EchoTherm™ Chilling/Heating Dry Baths
Description
Includes
Units come complete with bench-top power supply, instruction manual and AC line cord. Sample blocks and covers are available as accessories and are shown on the price list.
The IC22 2-Position Electronic Chilling/Heating Dry Bath is versatile, reliable and easy to use. Each heating/chilling station has its own Peltier module, and the entire unit has only two moving parts: the fans. Each plate can be set separately for any temperature within the range of unit without interaction between the plates. The IC22 has simple digital controls and a two-line alphanumeric digital display that is backlit for easy viewing. The display shows the target and actual temperatures of both plates.
- Energy-efficient Peltier design
- Digital control to 1°C from -10 to 100°C
- Two separate temperatures on separate plates
- Sample blocks for 0.2, 0.5 and 1.5mL centrifuge tubes, 96-well and 384-well assay plates, 2mL vials, various sizes of test tubes and more
- Control at room temperature
- Two countdown timers (up to 30 days each) with alarm
- RS232 I/O port for control or data collection
- Two data loggers
- Electronic calibration
- Compact size; small footprint on the bench
- Temperature Range on each plate: -10° to 100°C
- Readability: 1°C
- Accuracy: ±1°C
- Stability: ±1°C
- Sensor: Platinum RTD
- Controller Type: PID
- Uniformity: ±0.5°C
- Maintaining 14°C for ligation reactions
- Maintaining 17°C for storing oocytes
- Storing samples at ice bucket temperatures
- Incubate samples in 96-well assay plates, or in aluminum blocks holding 0.2, 0.5 or 1.5mL centrifuge tubes
- Cooling blood samples prior to coagulation testing
- Storing enzymes or DNA libraries at your work station
- Transporting samples from the field at exactly the right temperature
- Chilling or heating samples in the field
- Enzyme reactions and deactivations
- Freezing and storing most any small sample at your workstation
- PCR sample preparation
- Hybridizations
- General molecular biology applications
Compliance
UL, CSA and CE compliant
Warranty and Services
One year
Specifications
Specifications
Certifications/Compliance | UL, CSA, CE |
Temperature Range (Metric) | -10°C to +100°C |
Display | Digital |
Includes | Bench top power supply, instruction manual and AC line cord, sample blocks and covers |
Model | IC25 |
Run Time | 00:01 sec. to 30 days |
Temperature Accuracy | ±1°C |
Temperature Uniformity | ±0.5°C |
Warranty | 1 Year |
Width (English) | 6.5 in. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.