Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECHS1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP296977JF549
Description
ECHS1 Polyclonal antibody specifically detects ECHS1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ECHS1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 4.2.1, enoyl CoA hydratase, short chain, 1, mitochondrial, enoyl Coenzyme A hydratase, short chain, 1, mitochondrial, Enoyl-CoA hydratase 1, enoyl-CoA hydratase, mitochondrial, SCEHEC 4.2.1.17, Short-chain enoyl-CoA hydratase | |
Produced in rabbits immunized with purified, recombinant Human ECHS1 (rh ECHS1; AAH08906.1; 28A-290Q) | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
1892 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction