Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECHS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596259 Supplier Invitrogen™ Supplier No. PA596259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASGANFEYII AEKRGKNNTV GLIQLNRPKA LNALCDGLID ELNQALKTFE EDPAVGAIVL TGGDKAFAAG ADIKEMQNLS FQDCYSSKFL KHWDHLTQVK KPVIAAVNGY AFGGGCELAM MCDIIYAGEK AQFAQPEILI GTIPGAGGTQ RLTRAVGKSL AMEMVLTGDR ISAQDAKQAG LVSKICPVET LVEEAIQCAE KIASNSKIVV AMAKESVNAA FEMTLTEGSK LEKKLFYSTF ATDDRKEGMT AFVEKRKANF KDQ; Positive Samples: LO2, 293T, U-87MG, MCF7, 22Rv1, Mouse liver, Mouse kidney, Mouse heart, Rat liver; Cellular Location: Mitochondrion matrix.

The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECHS1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Echs1
Gene Accession No. P14604, P30084, Q8BH95
Gene Alias C80529; ECHS1; ECHS1D; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain 1; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; enoyl-CoA hydratase 1; Enoyl-CoA hydratase short chain 1 mitochondrial; enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain 1; Enoyl-CoA hydratase, short chain 1, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; mitochondrial short-chain enoyl-coenzyme A hydratase 1; SCEH; short chain enoyl-CoA hydratase; short-chain enoyl-CoA hydratase
Gene Symbols Echs1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-290 of human ECHS1 (NP_0040833).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140547, 1892, 93747
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.