Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECHS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597687
Description
ECHS1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.
Specifications
ECHS1 | |
Polyclonal | |
Unconjugated | |
Echs1 | |
C80529; ECHS1; ECHS1D; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain 1; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; enoyl-CoA hydratase 1; Enoyl-CoA hydratase short chain 1 mitochondrial; enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain 1; Enoyl-CoA hydratase, short chain 1, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; mitochondrial short-chain enoyl-coenzyme A hydratase 1; SCEH; short chain enoyl-CoA hydratase; short-chain enoyl-CoA hydratase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140547, 1892, 93747 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14604, P30084, Q8BH95 | |
Echs1 | |
Recombinant Human Enoyl-CoA hydratase, mitochondrial protein (28-290AA). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction