Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECM-1/Secretory Component P85 Antibody (030), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289452DL594
Description
ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
ECM-1/Secretory Component P85 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
30 | |
Sandwich ELISA Detection | |
ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ECM-1/Secretory Component P85 (NP_004416.1; Met1-Glu540). | |
0.1 mL | |
Extracellular Matrix, Immunology | |
1893 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction