Learn More
MilliporeSigma™ ECM Cell Culture Optimization Array (Colorimetric, 96-Wells)
Shop All MilliporeSigma ProductsDescription
Includes
- ECM Gradient Array Plate: One 96-well plate with 12 strips
- 30%BSA: One vial - 0.5mL
- Cell Stain Solution: One bottle - 20mL
- Extraction Buffer: One bottle - 20mL
- Assay Buffer: One bottle - 100mL
The 96-well, colorimetric array provides a multiparametric assay that allows researchers to:
- Quickly identify ECM(s) that best promote cell-type specific adhesion and
- Pinpoint the optimal ECM concentration required to promote maximum adhesion for a particular cell type
Specifications
Specifications
| Content And Storage | The ECM Gradient Array Plate should be stored in the foil pouch at -70°C, and used within three months of the date of receipt. The remaining kit components should be stored at 2° to 8°C |
| Detection Method | Colorimetric |
| For Use With (Application) | Quickly identify the best ECM protein for cell culture environment and pinpoint the concentration needed to achieve optimal cell growth conditions |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.