Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECM1 Recombinant Rabbit Monoclonal Antibody (001)
ECM1 Recombinant Rabbit Monoclonal Antibody (1)
Supplier: Thermo Scientific MA531112
Description

Specifications
ECM1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q61508 | |
ECM1 | |
Recombinant Mouse ECM1 protein (Met1-Glu559). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
001 | |
Unconjugated | |
ECM1 | |
AI663821; ECM1; Extracellular matrix protein 1; LOW QUALITY PROTEIN: extracellular matrix protein 1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD | |
Rabbit | |
Protein A | |
RUO | |
13601 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction