Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ecm29 (Ab Ecm29-3), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1052MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1052MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1052MI Supplier MilliporeSigma™ Supplier No. ABS1052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteasome-associated protein ECM29 homolog (UniProt Q5VYK3; also known as Ecm29, Extracellular mutant protein 29 homolog) is encoded by the ECM29 (also known as KIAA0368) gene (Gene ID 23392) in human. Ecm29 is a large adaptor protein found exclusively on the 26S enzyme to form a subset of 26S proteasomes (Ecm proteasomes), where it functions to link 26S proteasome to molecular motors, centrosome, endoplasmic reticulum, and endocytic vesicles. Ubiquitinated proteins containing K48- and K63-linked polyubiquitin chains accumulate in the centrosomes during M phase relative to S phase, shRNA-mediated Ecm29 knockdown is found to result in a decreased accumulation of ubiquitinated proteins in the centrosomes. Ecm29 downregulation is also reported to result in an elevated level of toll-like receptor 3 (TLR3) and enhanced TLR3 signaling as a result of impaired autophagy. Ecm29 is produced as a 1845-amino acid protein whose initiator methionine is removed posttranslationally to yiled the mature protein (a.a. 2-1845) composed almost entirely of Huntingtin-elongation factor 3-protein phosphatase 2A-Tor1 (HEAT) repeats, 27 in total (a.a. 34-1828).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ecm29 (Ab Ecm29-3)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene ECM29, KIAA0368
Gene Accession No. Q5VYK3
Host Species Rabbit
Immunogen Ovalbumin-conjugated linear peptide corresponding to the C-terminal sequence of human Ecm29.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) XP_005251907
Test Specificity This polyclonal antiserum (Ab Ecm29-3) stained both early and late endosomes in HeLa cells and detected the ∽205 kDa target band in HeLa cell lysate and in various mouse tissue homogenates. A high molecular weight Ecm29 isoform was also detected by this antiserum in mouse brain homogenate (Gorbea, C., et al., (2004). J. Biol. Chem. 2279(52):54849-54861).
Target Species Bovine
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.