Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECM29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584437 Supplier Invitrogen™ Supplier No. PA584437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVGLGTKGGC ASVIVSLTTQ CPQDLTPYSG KLMSALLSGL TDRNSVIQKS CAFAMGHLVR TSRDSSTEKL LQKLNGWYME KEEPIYKTSC ALTIHAIGRY SPDVLKNHAK EVL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

ECM29 appears to be one of three proteins which associate significantly with proteasomes. ECM29 has been shown to enhance the stability of the proteasome. ECM29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that ECM29 tethers the regulatory complex to the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECM29
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ECPAS
Gene Accession No. Q5VYK3
Gene Alias AI314180; AW558785; BB181316; ECM29; ECM29 homolog, proteasome accessory protein; Ecpas; expressed sequence AI314180; FLJ22036; homolog of yeast ECM29; KIAA0368; KIAA1962; mKIAA0368; Proteasome adapter and scaffold protein ECM29; Proteasome-associated protein ECM29 homolog; RP11-386D8.2
Gene Symbols ECPAS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ECPAS. Recombinant protein control fragment (Product #RP-103820).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.