Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ECMatrix™-411 E8 Laminin Subtrate
ECMatrix™-411 E8 Laminin Subtrate Xeno-free laminin-411 coating for the differentiation of pluripotent stem cells into endothelial cells
$696.13 - $1825.76
Specifications
Cell Type | Pluripotent stem cells into endothelial cells |
---|---|
Content And Storage | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
Form | Liquid |
Substrate | Laminin substrate |
Storage Requirements | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect them from light |
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
CC1621050MI
|
MilliporeSigma™
CC1621050UG |
6 x 175 μg |
Each for $1,825.76
|
|
|||||
CC162350UMI
|
MilliporeSigma™
CC162350UG |
2 x 175 μg |
Each for $696.13
|
|
|||||
Description
- Endothelial cells line the interior surface of blood and lymphatic vessels, forming an interface between circulating blood or lymph in the lumen and the rest of tissue/organ
- Laminin-411 binds to the integrin α6β1, located on the cell surface of pluripotent stem cells and induces endothelial cell differentiation
Specifications
Pluripotent stem cells into endothelial cells | |
Liquid | |
2° to 8°C, Avoid multiple freeze-thaw cycles and protect them from light |
2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light | |
Laminin substrate | |
0.5 mg/mL in PBS |
For research purposes only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title