Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ECMatrix™-511 E8 Laminin Substrate
Shop All MilliporeSigma Products
Click to view available options
Packaging:
100 mL
2 x 175 μg
6 x 175 μg
Description
- Human pluripotent stem cells (ES and iPS cells) express α6β1 as the major integrin species and can be maintained stably and expanded efficiently in feeder-free conditions on culture vessels coated with the binding partner laminin-511
- Animal-free, xeno-free format: Consistent from lot-to-lot with no prescreening required
- No plate precoating required: Save time by adding to media while passaging cells
- Supports single cell passaging without ROCKi: Great for CRISPR editing or clonal isolation
- Higher adhesion and growth rates: Get to your experiments faster
- Easy to handle: No chilling of cell culture consumables required

Specifications
Specifications
Cell Type | Pluripotent stem cell cultures, CHO-S derived |
Form | Liquid |
Substrate | Laminin |
Includes | 100 mL ECMatrix™-511 E8 Laminin Substrate (expressed in CHO-S cells) |
Packaging | 100 mL |
Storage Requirements | Store 2°C to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
Substrate Quantity | 1.6 μg/mL in recombinant HAS/PBS |
For research purposes only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction