Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eco1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Eco1 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Eco1 Polyclonal specifically detects Eco1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Eco1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
CTF7 homolog 1, EC 2.3.1.-, ECO1 homolog 1, EFO1CTF, EFO1p, ESO1 homolog 1, ESO1A930014I12Rik, Establishment factor-like protein 1, Establishment of cohesion 1 homolog 1, establishment of cohesion 1 homolog 1 (S. cerevisiae), hEFO1, KIAA1911ECO1, MGC105022, N-acetyltransferase ESCO1 | |
ESCO1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
DNA Repair, Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
114799 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ERAPENCHLANEIKPSDPPLDNQMKHSFDSASNKNFSQCLESKLENSPVENVTAASTLLSQAKIDTGENKFPGSAPQQHSILSNQTSKSSDNRETPRNHSLPKCNSHLEI | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title