Learn More
Sigma Aldrich Ecogenius PLA 3D printing filament
Catalog No. 503125895 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-312-5895 1 Ea.
1 options
Catalog No. 50-312-5895 Supplier Sigma Aldrich Supplier No. TRD3D00151EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
ParametersExtruder (nozzle) temperature: 195 +/-10°CBed temperature: cold to 80°CPrint speed: 60-80 mm/sFan: 50-100 %Minimum nozzle diameter: 0.3 mmPlate materials: glass, blue tape, etc.
Specifications
Quantity 1 Ea.