Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Economy Steel Tuning Fork
SCI_ED

Catalog No. S24891
Click to view available options
Frequency:
256 Hz
288 Hz
320 Hz
341 Hz
384 Hz
426 Hz
440 Hz
480 Hz
512 Hz

Economy steel tuning forks are designed for sound wave lessons and exploration

  • Chrome plated steel tuning fork
  • Economy priced
  • Frequency within ±5%

Specifications

Product Type Tuning Fork
Material Chrome-plated Steel
For Use With (Application) Science Experiments
Frequency 384 Hz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.