Learn More
Description
- Protect against groundwater contamination, using post-industrial material
- Robust pallets withstand the toughest environments for superior spill control
- Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
- Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
- Oversized leakproof sumps meet or exceed regulations
- Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
- Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
- Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
- Square pallet may be equipped with an optional ramp
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
Specifications
Specifications
| Certifications/Compliance | SPCC, EPA, NFPA, U (NFPA), meets fire codes |
| Color | Black |
| Capacity (English) | 66 gal. |
| Capacity (Metric) | 250 L |
| Material | Polyethylene |
| Type | Ecopolyblend Spill Control Pallet |
| For Use With (Application) | Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination |
| For Use With (Equipment) | Drums |
| Holds | 2 Drums |
| Includes | Without drain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

